Assay Details
| Gene Symbol: ttll-12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063589.6 | NP_495990.1 | 3 - 4 | 991 | 78 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ttll-12| Gene Symbol: ttll-12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_063589.6 | NP_495990.1 | 3 - 4 | 991 | 78 | |
Set Membership: |
|