Assay Details
| Gene Symbol: W03C9.5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_064149.6 | NP_496550.1 | 2 - 3 | 122 | 84 | |
| Gene Symbol: rab-7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_064148.8 | NP_496549.1 | 2 - 3 | 116 | 84 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_064148.8 transcripts.
Set Membership: |
|

W03C9.5
Genomic Map