Assay Details
| Gene Symbol: C29F9.6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_064743.4 | NP_497144.3 | 3 - 4 | 399 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

C29F9.6| Gene Symbol: C29F9.6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_064743.4 | NP_497144.3 | 3 - 4 | 399 | 63 | |
Set Membership: |
|