Assay Details
| Gene Symbol: pxd-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065403.6 | NP_497804.2 | 8 - 9 | 1468 | 95 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

pxd-1| Gene Symbol: pxd-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065403.6 | NP_497804.2 | 8 - 9 | 1468 | 95 | |
Set Membership: |
|