Assay Details
| Gene Symbol: ocr-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_068771.7 | NP_501172.2 | 9 - 10 | 2168 | 133 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ocr-4| Gene Symbol: ocr-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_068771.7 | NP_501172.2 | 9 - 10 | 2168 | 133 | |
Set Membership: |
|