Assay Details
| Gene Symbol: efa-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001129346.3 | NP_001122818.1 | 5 - 6 | 894 | 65 | |
| NM_001268723.3 | NP_001255652.1 | 3 - 4 | 496 | 65 | ||
| NM_070015.6 | NP_502416.1 | 7 - 8 | 1105 | 65 | ||
| NM_070016.6 | NP_502417.1 | 7 - 8 | 1105 | 65 | ||
| GenBank mRNA | EF473217.1 | - | 6 - 7 | 1048 | 65 | |
| EF473218.1 | - | 5 - 6 | 889 | 65 | ||
More Information
Set Membership: |
|

efa-6
Genomic Map