Assay Details
Gene Symbol: efa-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129346.3 | NP_001122818.1 | 6 - 7 | 1077 | 141 | |
NM_001268723.3 | NP_001255652.1 | 4 - 5 | 679 | 141 | ||
NM_070015.6 | NP_502416.1 | 8 - 9 | 1288 | 141 | ||
NM_070016.6 | NP_502417.1 | 8 - 9 | 1288 | 141 | ||
GenBank mRNA | EF473217.1 | - | 7 - 8 | 1231 | 141 | |
EF473218.1 | - | 6 - 7 | 1072 | 141 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
