Assay Details
| Gene Symbol: sos-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001373020.3 | NP_001360576.1 | 7 - 8 | 1505 | 134 | |
| NM_071834.5 | NP_504235.2 | 7 - 8 | 1517 | 134 | ||
More Information
Set Membership: |
|

sos-1
Genomic Map