Assay Details
| Gene Symbol: grl-8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_072306.2 | NP_504707.1 | 2 - 3 | 369 | 155 | |
| Gene Symbol: grl-9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_072305.5 | NP_504706.1 | 2 - 3 | 358 | 155 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_072305.5 transcripts.
Set Membership: |
|

grl-8
Genomic Map