Assay Details
Gene Symbol: gna-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_073253.7 | NP_505654.1 | 1 | 456 | 70 | |
GenBank mRNA | AB017628.1 | - | 1 | 456 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
