Assay Details
| Gene Symbol: Y87G2A.18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026663.4 | NP_001021834.1 | 1 - 2 | 60 | 78 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Y87G2A.18| Gene Symbol: Y87G2A.18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026663.4 | NP_001021834.1 | 1 - 2 | 60 | 78 | |
Set Membership: |
|