Assay Details
| Gene Symbol: RTN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007131.1 | NP_001007132.1 | 5 - 6 | 623 | 84 | |
| XM_022421749.1 | XP_022277457.1 | 7 - 8 | 2037 | 84 | ||
More Information
Set Membership: |
|

RTN1
Genomic Map