Assay Details
| Gene Symbol: CCL17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003051.1 | NP_001003051.1 | 2 - 3 | 186 | 76 | |
| XM_005617215.3 | XP_005617272.1 | 4 - 5 | 4087 | 76 | ||
| GenBank mRNA | AB054642.1 | - | 2 - 3 | 186 | 76 | |
More Information
Set Membership: |
|

CCL17
Genomic Map