Assay Details
Gene Symbol: RPL19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001313782.1 | NP_001300711.1 | 1 | 335 | 85 | |
GenBank mRNA | AJ388522.1 | - | 1 | 144 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
