Assay Details
| Gene Symbol: TSPAN31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014117197.2 | XP_013972672.1 | 215 | 64 | ||
| XM_531651.6 | XP_531651.1 | 206 | 64 | |||
| GenBank mRNA | AJ388529.1 | - | 164 | 64 | ||
More Information
Set Membership: |
|

TSPAN31
Genomic Map