Assay Details
| Gene Symbol: TLR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005264.3 | NP_001005264.2 | 1 | 2175 | 69 | |
| XM_005628829.3 | XP_005628886.1 | 2 | 2220 | 69 | ||
| XM_022427656.1 | XP_022283364.1 | 2 | 2270 | 69 | ||
| XM_022427657.1 | XP_022283365.1 | 2 | 2248 | 69 | ||
| GenBank mRNA | AJ630583.1 | - | 1 | 202 | 69 | |
| EU487534.1 | - | 1 | 2175 | 69 | ||
More Information
Set Membership: |
|

TLR2
Genomic Map