Assay Details
| Gene Symbol: NGF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194950.1 | NP_001181879.1 | 1 | 546 | 124 | |
| GenBank mRNA | AY800386.1 | - | 1 | 427 | 124 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: NGF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194950.1 | NP_001181879.1 | 1 | 546 | 124 | |
| GenBank mRNA | AY800386.1 | - | 1 | 427 | 124 | |
Set Membership: |
|