Assay Details
| Gene Symbol: TPMT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003015.2 | NP_001003015.1 | 3 - 4 | 357 | 74 | |
| XM_005639955.3 | XP_005640012.1 | 4 - 5 | 418 | 74 | ||
| XM_005639956.3 | XP_005640013.1 | 3 - 4 | 343 | 74 | ||
| XM_014110479.2 | XP_013965954.1 | 4 - 5 | 588 | 74 | ||
| XM_022414374.1 | XP_022270082.1 | 4 - 5 | 418 | 74 | ||
| GenBank mRNA | AY057077.1 | - | 3 - 4 | 357 | 74 | |
| AY057078.1 | - | 2 - 3 | 237 | 74 | ||
More Information
Set Membership: |
|

TPMT
Genomic Map