Assay Details
| Gene Symbol: TFF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002991.1 | NP_001002991.1 | 2 - 3 | 239 | 65 | |
| GenBank mRNA | AY264843.1 | - | 2 - 3 | 239 | 65 | |
| EF591860.1 | - | 2 - 3 | 223 | 65 | ||
| EF591861.1 | - | 2 - 3 | 223 | 65 | ||
| EF641293.1 | - | 2 - 3 | 223 | 65 | ||
More Information
Set Membership: |
|

TFF2
Genomic Map