Assay Details
| Gene Symbol: TFF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002991.1 | NP_001002991.1 | 3 - 4 | 381 | 81 | |
| GenBank mRNA | AY264843.1 | - | 3 - 4 | 381 | 81 | |
| EF591861.1 | - | 3 - 4 | 365 | 81 | ||
More Information
Set Membership: |
|

TFF2
Genomic Map