Assay Details
| Gene Symbol: HTR2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024633.1 | NP_001019804.1 | 2 - 3 | 545 | 102 | |
| GenBank mRNA | AY832859.1 | - | 2 - 3 | 545 | 102 | |
More Information
Set Membership: |
|

HTR2B
Genomic Map