Assay Details
| Gene Symbol: VCAM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003298.1 | NP_001003298.1 | 3 - 4 | 755 | 91 | |
| XM_005621302.3 | XP_005621359.2 | 3 - 4 | 824 | 91 | ||
| XM_005621304.2 | XP_005621361.1 | 3 - 4 | 676 | 91 | ||
| XM_005621305.3 | XP_005621362.1 | 3 - 4 | 618 | 91 | ||
| GenBank mRNA | U32086.1 | - | 3 - 4 | 755 | 91 | |
More Information
Set Membership: |
|

VCAM1
Genomic Map