Assay Details
| Gene Symbol: ZNF432 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005616216.3 | XP_005616273.1 | 2 - 3 | 396 | 75 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005616216.3 transcripts.
Set Membership: |
|

ZNF432
Genomic Map