Assay Details
| Gene Symbol: OLFML1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005633615.2 | XP_005633672.1 | 2 - 3 | 503 | 100 | |
| XM_534043.6 | XP_534043.2 | 2 - 3 | 980 | 100 | ||
More Information
Set Membership: |
|

OLFML1
Genomic Map