Assay Details
Gene Symbol: PPAN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204439.1 | NP_001191368.1 | 6 - 7 | 610 | 59 | |
GenBank mRNA | BM537261.1 | - | 4 - 5 | 350 | 59 | |
DN371251.1 | - | 6 - 7 | 607 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
