Assay Details
Gene Symbol: POLR1D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014107409.2 | XP_013962884.1 | 198 | 104 | ||
XM_022409965.1 | XP_022265673.1 | 368 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
