Assay Details
Gene Symbol: NDUFV3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005638943.3 | XP_005639000.1 | 1 - 2 | 148 | 88 | |
XM_535598.6 | XP_535598.3 | 1 - 2 | 148 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
