Assay Details
| Gene Symbol: ZNF300 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022417801.1 | XP_022273509.1 | 3 - 4 | 521 | 77 | |
| XM_022417802.1 | XP_022273510.1 | 3 - 4 | 326 | 77 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_022417801.1, XM_022417802.1 transcripts.
Set Membership: |
|

ZNF300
Genomic Map