Assay Details
| Gene Symbol: ZNF300 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022417801.1 | XP_022273509.1 | 4 - 5 | 653 | 150 | |
| XM_022417802.1 | XP_022273510.1 | 4 - 5 | 458 | 150 | ||
More Information
Set Membership: |
|

ZNF300
Genomic Map