Assay Details
| Gene Symbol: ZNF330 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005631775.3 | XP_005631832.1 | 651 | 67 | ||
| XM_014121485.2 | XP_013976960.1 | 672 | 67 | |||
| XM_533282.7 | XP_533282.2 | 671 | 67 | |||
More Information
Set Membership: |
|

ZNF330
Genomic Map