Assay Details
| Gene Symbol: NAA38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022418536.1 | XP_022274244.1 | 2 - 3 | 380 | 83 | |
| XM_022418537.1 | XP_022274245.1 | 2 - 3 | 877 | 83 | ||
More Information
Set Membership: |
|

NAA38
Genomic Map