Assay Details
| Gene Symbol: POC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022416776.1 | XP_022272484.1 | 970 | 97 | ||
| XM_022416777.1 | XP_022272485.1 | 834 | 97 | |||
| XM_022416778.1 | XP_022272486.1 | 970 | 97 | |||
| XM_022416779.1 | XP_022272487.1 | 742 | 97 | |||
| XM_022416780.1 | XP_022272488.1 | 880 | 97 | |||
| XM_022416781.1 | XP_022272489.1 | 877 | 97 | |||
| XM_022416783.1 | XP_022272491.1 | 970 | 97 | |||
| XM_022416784.1 | XP_022272492.1 | 970 | 97 | |||
| XM_536320.6 | XP_536320.3 | 967 | 97 | |||
More Information
Set Membership: |
|

POC5
Genomic Map