Assay Details
| Gene Symbol: TEFM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005624831.3 | XP_005624888.1 | 1 - 2 | 148 | 61 | |
| XM_537735.6 | XP_537735.1 | 1 - 2 | 148 | 61 | ||
More Information
Set Membership: |
|

TEFM
Genomic Map