Assay Details
Gene Symbol: TIMM22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_537758.6 | XP_537758.1 | 3 - 4 | 839 | 55 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
