Assay Details
| Gene Symbol: TMEM219 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005621341.3 | XP_005621398.1 | 3 - 4 | 534 | 116 | |
| XM_005621342.2 | XP_005621399.1 | 5 - 6 | 978 | 116 | ||
| XM_014114490.2 | XP_013969965.1 | 2 - 3 | 1329 | 116 | ||
| XM_022419879.1 | XP_022275587.1 | 2 - 3 | 1333 | 116 | ||
| XM_022419880.1 | XP_022275588.1 | 3 - 4 | 570 | 116 | ||
| XM_022419881.1 | XP_022275589.1 | 4 - 5 | 793 | 116 | ||
| XM_022419883.1 | XP_022275591.1 | 2 - 3 | 408 | 116 | ||
| XM_536912.6 | XP_536912.2 | 3 - 4 | 521 | 116 | ||
More Information
Set Membership: |
|

TMEM219
Genomic Map