Assay Details
| Gene Symbol: TERF2IP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_536776.6 | XP_536776.2 | 2 - 3 | 930 | 125 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TERF2IP| Gene Symbol: TERF2IP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_536776.6 | XP_536776.2 | 2 - 3 | 930 | 125 | |
Set Membership: |
|