Assay Details
| Gene Symbol: GATAD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005628510.3 | XP_005628567.1 | 3 - 4 | 678 | 89 | |
| XM_022427339.1 | XP_022283047.1 | 6 - 7 | 877 | 89 | ||
| XM_539418.5 | XP_539418.3 | 4 - 5 | 736 | 89 | ||
More Information
Set Membership: |
|

GATAD1
Genomic Map