Assay Details
| Gene Symbol: HEBP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003638773.4 | XP_003638821.1 | 317 | 71 | ||
| XM_014112344.2 | XP_013967819.1 | 159 | 71 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: HEBP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003638773.4 | XP_003638821.1 | 317 | 71 | ||
| XM_014112344.2 | XP_013967819.1 | 159 | 71 | |||
Set Membership: |
|