Assay Details
| Gene Symbol: DLA-DMA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005627068.3 | XP_005627125.1 | 3 - 4 | 998 | 105 | |
| XM_014118041.2 | XP_013973516.1 | 2 - 3 | 417 | 105 | ||
| XM_014118042.2 | XP_013973517.1 | 2 - 3 | 394 | 105 | ||
More Information
Set Membership: |
|

DLA-DMA
Genomic Map