Assay Details
| Gene Symbol: SH3BP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005635907.3 | XP_005635964.1 | 2951 | 62 | ||
| XM_022409727.1 | XP_022265435.1 | 2837 | 62 | |||
| XM_543298.6 | XP_543298.2 | 2848 | 62 | |||
More Information
Set Membership: |
|

SH3BP4
Genomic Map