Assay Details
| Gene Symbol: ZSCAN16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022414590.1 | XP_022270298.1 | 3 - 4 | 727 | 75 | |
| XM_022414591.1 | XP_022270299.1 | 3 - 4 | 681 | 75 | ||
| XM_022414592.1 | XP_022270300.1 | 3 - 4 | 824 | 75 | ||
More Information
Set Membership: |
|

ZSCAN16
Genomic Map