Assay Details
| Gene Symbol: MMRN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005619114.3 | XP_005619171.1 | 2 - 3 | 369 | 59 | |
| XM_014112945.2 | XP_013968420.1 | 2 - 3 | 371 | 59 | ||
| XM_022417616.1 | XP_022273324.1 | 2 - 3 | 371 | 59 | ||
More Information
Set Membership: |
|

MMRN2
Genomic Map