Assay Details
| Gene Symbol: MCMDC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022412357.1 | XP_022268065.1 | 14 - 15 | 2039 | 130 | |
| XM_022412358.1 | XP_022268066.1 | 13 - 14 | 2057 | 130 | ||
| XM_022412359.1 | XP_022268067.1 | 14 - 15 | 2036 | 130 | ||
| XM_022412360.1 | XP_022268068.1 | 13 - 14 | 1991 | 130 | ||
| XM_544111.6 | XP_544111.3 | 14 - 15 | 2117 | 130 | ||
More Information
Set Membership: |
|

MCMDC2
Genomic Map