Assay Details
| Gene Symbol: DDIT4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_546156.6 | XP_546156.2 | 1 - 2 | 164 | 86 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_546156.6 transcripts.
Set Membership: |
|

DDIT4
Genomic Map