Assay Details
| Gene Symbol: OSGIN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005620648.3 | XP_005620705.2 | 6 - 7 | 755 | 59 | |
| XM_005620650.2 | XP_005620707.1 | 5 - 6 | 668 | 59 | ||
More Information
Set Membership: |
|

OSGIN1
Genomic Map