Assay Details
| Gene Symbol: KIAA0753 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005619873.2 | XP_005619930.1 | 14 - 15 | 2833 | 95 | |
| XM_005619876.2 | XP_005619933.1 | 14 - 15 | 2846 | 95 | ||
| XM_022418407.1 | XP_022274115.1 | 13 - 14 | 2756 | 95 | ||
| XM_022418408.1 | XP_022274116.1 | 12 - 13 | 1489 | 95 | ||
| XM_843538.4 | XP_848631.2 | 14 - 15 | 2845 | 95 | ||
More Information
Set Membership: |
|

KIAA0753
Genomic Map