Assay Details
| Gene Symbol: GTPBP8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003434098.4 | XP_003434146.1 | 3 - 4 | 595 | 91 | |
| XM_844347.5 | XP_849440.1 | 4 - 5 | 694 | 91 | ||
More Information
Set Membership: |
|

GTPBP8
Genomic Map