Assay Details
| Gene Symbol: HSD17B2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005620652.3 | XP_005620709.1 | 1 - 2 | 579 | 124 | |
| XM_005620653.3 | XP_005620710.1 | 1 - 2 | 580 | 124 | ||
| XM_022419125.1 | XP_022274833.1 | 1 - 2 | 586 | 124 | ||
| XM_546810.5 | XP_546810.3 | 1 - 2 | 576 | 124 | ||
More Information
Set Membership: |
|

HSD17B2
Genomic Map