Assay Details
Gene Symbol: ICMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_546748.6 | XP_546748.1 | 243 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
