Assay Details
| Gene Symbol: TEX35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022420996.1 | XP_022276704.1 | 4 - 5 | 440 | 111 | |
| XM_022420997.1 | XP_022276705.1 | 4 - 5 | 417 | 111 | ||
| XM_022420999.1 | XP_022276707.1 | 4 - 5 | 389 | 111 | ||
| XM_022421001.1 | XP_022276709.1 | 4 - 5 | 363 | 111 | ||
| XM_022421002.1 | XP_022276710.1 | 4 - 5 | 335 | 111 | ||
| XM_022421003.1 | XP_022276711.1 | 4 - 5 | 309 | 111 | ||
| XM_022421004.1 | XP_022276712.1 | 3 - 4 | 284 | 111 | ||
More Information
Set Membership: |
|

TEX35
Genomic Map